missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Antibody (5E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £307.00
Specifications
| Antigen | CYP1B1 |
|---|---|
| Clone | 5E2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648997
|
Novus Biologicals
NBP2-50190-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670808
|
Novus Biologicals
NBP2-50190 |
0.1 mL |
£307.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP1B1 Monoclonal antibody specifically detects CYP1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP1B1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| PBS | |
| 1545 | |
| IgG1 κ | |
| Protein A purified |
| 5E2 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase | |
| Ovalbumin-conjugated synthetic peptide - PENFDPARFLDKDGL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title