missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP1A1/CYP1A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101307
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytochrome P450 1A1/2.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. Other xenobiotic substrates for this enzyme include caffeine, aflatoxin B1, and acetaminophen. The transcript from this gene contains four Alu sequences flanked by direct repeats in the 3' untranslated region.
Specifications
| CYP1A1/CYP1A2 | |
| Polyclonal | |
| Unconjugated | |
| CYP1A2 | |
| AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP11; CP12; CYP1; Cyp1a1; Cyp1a-1; CYP1A2; Cyp1a-2; Cyp45c; Cypc45c; CYPD45; CYPIA1; CYPIA2; Cytochrome P(3)450; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P3 450; cytochrome P-448; cytochrome P450 1A1; Cytochrome P450 1A2; cytochrome P450 4; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily A member 2; cytochrome P450 family 1 subfamily a polypeptide 1; cytochrome P450 family 1 subfamily a polypeptide 2; cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily a, polypeptide 1; cytochrome P450, family 1, subfamily a, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; cytochrome P450-C; Cytochrome P-450d; Cytochrome P450-D; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; cytochrome P450-P1; Cytochrome P450-P2; cytochrome P450-P3; dioxin-inducible; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P3-450; P-448; P450 form 4; P450(PA); P450-1; P450-3; P450-C; P-450d; P450-D; P450DX; P450form6; P-450MC; P450MT2; RATCYPD45; xenobiotic monooxygenase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13076, 13077, 1543, 1544, 24296, 24297 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P00184, P00185, P00186, P04798, P04799, P05177 | |
| CYP1A1, CYP1A2 | |
| A synthesized peptide derived from human CYP1A1(Accession P04798), corresponding to amino acid residues I78-G128. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction