missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP1A1 Polyclonal Antibody, DyLight™ 488
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579126
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - Flow: K562 cell.
This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.
Specifications
| CYP1A1 | |
| Polyclonal | |
| DyLight 488 | |
| CYP1A1 | |
| AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; CP11; CYP; CYP1; Cyp1a1; Cyp1a-1; Cyp1a2; Cyp45c; Cypc45c; CYPIA1; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; cytochrome P450 1A1; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily a polypeptide 2; cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, family 1, subfamily a, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450-C; cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; Cytochrome P450-P1; dioxin-inducible; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P450; P450 form 6; P450-1; P450-C; P450DX; P450form6; P-450MC; P450MT2; P450-P1; xenobiotic monooxygenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1543 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P04798 | |
| CYP1A1 | |
| E.coli-derived human CYP1A1 recombinant protein (Position: H183-D320). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction