missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP11B2 Recombinant Rabbit Monoclonal Antibody (K01_3J89)
GREENER_CHOICE

Product Code. 30827857
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827857 100 μL
30829592 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827857 Supplier Invitrogen™ Supplier No. MA564317

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CYP11B2 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the mitochondrial inner membrane and is involved in the conversion of progesterone to cortisol in the adrenal cortex. Mutations in this gene cause congenital adrenal hyperplasia due to 11-beta-hydroxylase deficiency. Transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11B2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3J89
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene CYP11B2
Gene Accession No. P19099
Gene Alias ALDOS; Aldosterone synthase; aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; Cp45as; Cpn2; Cyp11b; CYP11B2; Cyp11b-2; Cyp11b3; CYP11BL; CYPXIB2; CYPXIB3; cytochrome P450 11B2, mitochondrial; cytochrome P450 11B3, mitochondrial; cytochrome P450 family 11 subfamily B member 2; Cytochrome P450 subfamily XIB polypeptide 2 (aldosterone synthase); cytochrome P450, family 11, subfamily b, polypeptide 2; cytochrome P450, subfamily 11B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450, subfamily XIB, polypeptide 2 (aldosterone synthase); cytochrome P-450Aldo; cytochrome P450-Aldo-1; cytochrome P450-Aldo-2; cytochrome P450C11; Cytochrome P-450C18; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; P450aldo; P450-Aldo-1; P450-Aldo-2; P450C 18; P-450C 18; P450C18; P-450C18; RNCP45AS; steroid 11-beta/18-hydroxylase; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-beta-monooxygenase; Steroid 18-hydroxylase; steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo; steroid-11-beta-hydroxylase
Gene Symbols CYP11B2
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1585
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.