missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP11B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | CYP11B2 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646806
|
Novus Biologicals
NBP2-49267-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683169
|
Novus Biologicals
NBP2-49267 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP11B2 Polyclonal antibody specifically detects CYP11B2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CYP11B2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2), 40% Glycerol | |
| 1585 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ALDOSCYP11B, Aldosterone synthase, Aldosterone-synthesizing enzyme, CPN2, CYP11BL, CYPXIB2, cytochrome P450 11B2, mitochondrial, cytochrome P450, family 11, subfamily B, polypeptide 2, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2, Cytochrome P-450Aldo, Cytochrome P-450C18, EC 1.14.15, EC 1.14.15.4, EC 1.14.15.5, mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2, P450aldo, P450C18, P-450C18, steroid 11-beta/18-hydroxylase, steroid 11-beta-monooxygenase, Steroid 18-hydroxylase, steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VQQILRQESLAAAASISEHPQKATTELPLLRAALKETLR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title