missing translation for 'onlineSavingsMsg'
Learn More

CYP11B1 Antibody, Novus Biologicals™

Product Code. 18337557 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337557 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337557 Supplier Novus Biologicals Supplier No. H00001584D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP11B1 Polyclonal antibody specifically detects CYP11B1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11B1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH96285.1
Gene Alias CPN1, CYPXIB1, cytochrome P450 11B1, mitochondrial, cytochrome p450 XIB1, cytochrome P450, family 11, subfamily B, polypeptide 1, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1, Cytochrome P-450c11, Cytochrome P450C11, EC 1.14.15, EC 1.14.15.4, FHICYP11B, FLJ36771, P450C11DKFZp686B05283, S11BH, Steroid 11-beta-hydroxylase, steroid 11-beta-monooxygenase
Host Species Rabbit
Immunogen CYP11B1 (AAH96285.1, 1 a.a. - 574 a.a.) full-length human protein. MALRAKAEVCMAVPWLSLQRAQALGTRAARVPRTVLPFEAMPRRPGNRWLRLLQIWREQGYEDLHLEVHQTFQELGPIFRSRHSASFGRWGRSAARAGLWRCQGRGWCRANPSSLQRGQDSEALKYDLGGAGMVCVMLPEDVEKLQQVDSLHPHRMSLEPWVAYRQHRGHKCGVFLLNVADRGNSSPPFPGGIHGAPTHSGCRNGPEWRFNRLRLNPEVLSPNAVQRFLPMVDAVARDFSQALKKKVLQNARGSLTLDVQPSIFHYTIEASNLALFGERLGLVGHSPSSASLNFLHALEVMFKSTVQLMFMPRSLSRWTSPKVWKEHFEAWDCIFQYGDNCIQKIYQELAFSRPQQYTSIVAELLLNAELSPDAIKANSMELTAGSVDTTVFPLLMTLFELARNPNVQQALRQESLAAAASISEHPQKATTELPLLRAALKETLRLYPVGLFLERVASSDLVLQNYHIPAGTLVRVFLYSLGRNPALFPRPERYNPQRWLDIRGSGRNFYHVPFGFGMRQCLGRRLAEAEMLLLLHHVLKHLQVETLTQEDIKMVYSFILRPSMFPLLTFRAIN
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1584
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.