missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Cynomolgus Monkey MD2 (NP_001274662) VersaClone cDNA

Produktkode 18715479
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
Förpackningsstorlek:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Förpackningsstorlek
18715479 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 18715479 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' RDC2256
  • ONLINE EXCLUSIVE PRICE
    £301.00
    £284.55 / 10µg
    Save £16.45
    5% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Tilføj til indkøbskurv
Tilføj til indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

R&D Systems™ Cynomolgus Monkey MD2 (NP_001274662) VersaClone cDNA associates with toll-like receptor 4 on the cell surface and confers responsiveness to lipopolysaccyaride (LPS).

The Cynomolgus Monkey MD2 (NP_001274662) VersaClone cDNA clone contains the complete ORF for MD-2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MD-2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Tekniske data

Gene cynoLY96
Accession Number NP_001274662
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 496bp
pH 8.5
Gene ID (Entrez) 23643
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.