missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cynomolgus Monkey IL-23R Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB106241-SP
This item is not returnable.
View return policy
Description
IL-23R Monoclonal antibody specifically detects IL-23R in Cynomolgus Monkey samples. It is validated for Immunohistochemistry
Specifications
| IL-23R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Human embryonic kidney cell HEK293-derived cynomolgus monkey IL-23R, Gly24-Asp353, Accession # XP_005543141.1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 1032646 | |
| Immunohistochemistry 5-25 ug/mL | |
| XP_005543141.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 149233 | |
| Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction