missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cynomolgus Monkey CD2 (NP_001270706) VersaClone cDNA
Description
- CD2 is a type I glycoprotein belonging to the Ig superfamily and is expressed on T, NK, B, and some antigen presenting cells.
- CD2 binds CD58 on antigen presenting cells to induce co-stimulating signals in T cells.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoCD2 |
| Accession Number | NP_001270706 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1069 bp |
| Gene ID (Entrez) | 914 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?