missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cynomolgus Monkey CCL4/MIP-1 beta (Unique) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- CCL4 is a member of the CC or beta chemokine subfamily.
- CCL4 is expressed primarily by T cells, B cells, and monocytes after antigen or mitogen stimulation.
- The functional receptor for CCL4 has been identified as CCR5.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoLOC102117861 |
| Accession Number | unique |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 292 bp |
| Gene ID (Entrez) | 6351 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction