missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cyno Monkey LIFR alpha (XP_005556818.1) VersaClone cDNA

Product Code. 18742987
Change view
Click to view available options
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18742987 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18742987 Supplier R&D Systems™ Supplier No. RDC3156

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene LIFR alpha
Accession Number XP_005556818.1
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 3307 bp
Gene ID (Entrez) 3977
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF R alpha, LIF R beta, LIF receptor, LIFR, LIF-R, LIFRa, SJS2, STWS, SWS
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.