missing translation for 'onlineSavingsMsg'
Learn More

Cynaropicrin, MedChemExpress

Product Code. 16489288
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489288 10 mg 10mg
16421718 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489288 Supplier Medchem Express Supplier No. HYN2350
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cynaropicrin is a sesquiterpene lactone which can inhibit tumor necrosis factor (TNF-α) release with IC50s of 8.24 and 3.18 μM for murine and human macrophage cells, respectively. Cynaropicrin also inhibits the increase of cartilage degradation factor (MMP13) and suppresses NF-κB signaling.

Cynaropicrin is a sesquiterpene lactone which can inhibit tumor necrosis factor (TNF-α) release with IC50s of 8.24 and 3.18 μM for murine and human macrophage cells, respectively. Cynaropicrin also inhibits the increase of cartilage degradation factor (MMP13) and suppresses NF-κB signaling.

Specifications

Chemical Name or Material Cynaropicrin
CAS 35730-78-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C19H22O6
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (144.35 mM)
SMILES C=C([C@H]1O)[C@]([C@@]2([H])C1)([H])[C@](OC3=O)([H])[C@@](C3=C)([H])[C@@H](OC(C(CO)=C)=O)CC2=C
Molecular Weight (g/mol) 346.37
Formula Weight 346.37
Percent Purity 97.16%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.