missing translation for 'onlineSavingsMsg'
Learn More

Cynarin, MedChemExpress

Product Code. 16428958
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16428958

Brand: Medchem Express HYN0359

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.

Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.

Specifications

Chemical Name or Material Cynarin
Physical Form Solid
CAS 30964-13-7
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H24O12
For Use With (Application) COVID-19-anti-virus
Synonym Cynarine
Solubility Information DMSO : ≥ 23 mg/mL (44.53 mM)
SMILES O=C([C@]1(OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.