missing translation for 'onlineSavingsMsg'
Learn More

Cynarin, MedChemExpress

Product Code. 16421388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421388 5 mg 5mg
16473856 25 mg 25mg
16428958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16421388 Supplier Medchem Express Supplier No. HYN03595mg
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.

Cynarin is an antichoke agent with a variety of biological activities including antioxidant, antihistamic and antiviral activities.

Specifications

Chemical Name or Material Cynarin
Physical Form Solid
CAS 30964-13-7
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H24O12
For Use With (Application) COVID-19-anti-virus
Synonym Cynarine
Solubility Information DMSO : ≥ 23 mg/mL (44.53 mM)
SMILES O=C([C@]1(OC(/C=C/C2=CC=C(O)C(O)=C2)=O)C[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@@H](O)[C@H](O)C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.