missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 5442 hydrochloride
GREENER_CHOICE

Product Code. 18005822
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18005822

Brand: Tocris Bioscience™ 3601/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective S1P1 receptor agonist

CYM 5442 hydrochloride is a potent and selective S1P1 agonist in vitro (EC50 = 1.35 nM). Activates S1P1-mediated p42/p44 MAPK phosphorylation in CHO-K1 cells transfected with S1P1. Induces acute lymphopenia in mice. Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1783987-80-3
Quantity 10 mg
Product Type CYM 5442 hydrochloride
Purity >98%
Molecular Weight (g/mol) 445.94
Inhibitors CYM 5442 hydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.