missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50769
GREENER_CHOICE

Product Code. 30001994
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001994

Brand: Tocris Bioscience™ 4948/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novel non-peptide NPBWR1 antagonist

Novel non-peptide antagonist of neuropeptide W/B receptor 1 (NPBWR1, GPR7) (IC50 = 0.12 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421365-63-0
Molecular Formula C24H17ClN2O3

Specifications

Quantity 50 mg
Formula Weight 416.86
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 416.861
Chemical Name or Material CYM 50769
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.