missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50358 hydrochloride
GREENER_CHOICE

Product Code. 30002053
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002053 10 mg 10mg
30002059 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002053 Supplier Tocris Bioscience™ Supplier No. 4679/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective S1P4 antagonist

Potent S1P4 antagonist (IC50 = 25 nM). Displays selectivity for S1P4 against S1P1, S1P2, S1P3 and S1P5 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H18Cl2N2O2·HCl

Specifications

Quantity 10 mg
Formula Weight 425.74
Purity >98%
Chemical Name or Material CYM 50358 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.