Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polystyrene
Product Code. 18048031
Change view
Click to view available options
Capacity (Metric):
10 mL
15 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 700
Pack of 750
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility unitSize
18048031 10 mL Non-sterile Pack of 1000
18476898 10 mL Sterile Pack of 750
18008041 15 mL Non-sterile Pack of 750
18466898 15 mL Sterile Pack of 700
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18048031 Supplier Kartell Supplier No. 8820000
Only null left
Add to basket
Add to basket
Cylindrical test tubes equipped with a tight screw cap.
  • Test tubes are intended to contain blood and biological liquids to perform laboratory tests
Specifications
Closure Material Polypropylene
Capacity (Metric) 10 mL
Length (Metric) 100 mm
Material Polystyrene
Shape Cylindrical
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No
Closure Type Screw Cap
Product Type Test Tube