Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polypropylene
Product Code. 18078031
Click to view available options
Closure Color:
Red
Yellow
Sterility:
Non-sterile
Unit Size:
Pack of 1000
Product Code. 18078031
Brand: Kartell 8830306
Only null left
Add to basket
Add to basket
Test tubes with quick closure screw caps (PE). Leak-proof, and suitable for centrifugation. The design of the cap avoids any contamination due to drops of liquid on the inner surface.
  • For laboratory use
  • For collection and evaluation of samples
  • Caps are not autoclavable
Specifications
Closure Color Yellow
Color Clear
Capacity (Metric) 15 mL
Length (Metric) 100 mm
Material Polypropylene
Shape Cylindrical
Disposable Single-use
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Closure Type Screw Cap
Product Type Centrifuge Tube