missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ All-Purpose Cylindrical Stirring Bars

Product Code. 10274801
Change view
Click to view available options
Length (Metric):
6 mm
11.68 mm
14.80 mm
19.60 mm
24.61 mm
29.88 mm
40 mm
49.90 mm
60.16 mm
Diameter (Metric):
3 mm
4.57 mm
4.66 mm
6 mm
6.16 mm
7.87 mm
8 mm
9.62 mm
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
10274801 24.61 mm 6 mm Pack of 10
10192471 6 mm 3 mm Pack of 10
10544471 14.80 mm 4.57 mm Pack of 10
10192661 29.88 mm 6.16 mm Pack of 10
10152521 11.68 mm 4.66 mm Pack of 10
10306021 49.90 mm 7.87 mm Pack of 10
10355921 19.60 mm 6 mm Pack of 10
10524851 40 mm 8 mm Pack of 10
10192611 60.16 mm 9.62 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10274801 Supplier Kartell™ Supplier No. 0075700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features rounded shape with smooth walls.

  • PTFE-coated magnet
  • Magnetic field causes stirring bar to spin very quickly
  • Non-stick, non-reactive and hydrophobic
  • Highly resistant to thermo-shock
  • Suitable for foodstuff

Specifications

Description Magnet PTFE Coated
Diameter (Metric) 6 mm
Length (Metric) 24.61 mm
Quantity 10/Pk.
Shape Cylinder, Sphere
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.