missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cylicin 1 Antibody [Unconjugated], Novus Biologicals™
Rabbit Polyclonal Antibody
£321.00 - £503.00
Specifications
| Antigen | Cylicin 1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279503
|
Novus Biologicals
NBP3-43686-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279450
|
Novus Biologicals
NBP3-43686-100ul |
100 μL |
£503.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cylicin 1 Polyclonal antibody specifically detects Cylicin 1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| Cylicin 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CYCL1, CYL, CYL1, cylicin 1, cylicin I, cylicin, basic protein of sperm head cytoskeleton 1, cylicin-1, multiple-band polypeptide I | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GAKKKIDESDGTSANSKMEGLESKRGFRMSSKKTTFNEKGEKASTGRVPPSREKPPLPACEPSLPSPKVRRLCWCKMPPPPPKPRYAP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 1538 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title