missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYLD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583991
This item is not returnable.
View return policy
Description
Immunogen sequence: PPLEINSRVS LKVGETIESG TVIFCDVLPG KESLGYFVGV DMDNPIGNWD GRFDGVQLCS FACVESTILL HINDIIPESV TQER Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 95%.
CYLD is a 956 aa, cytoplasmic, deubiquitinating enzyme belonging to the ubiquitin carboxy-terminal hydrolases (UCH) family of proteins with three cytoskeletal-associated protein-glycine-conserved (CAP-GLY) domains, a proline rich region, a SH3 binding domain and a sequence homology to catalytic domain of UCH. CYLD is identified as a tumor suppressor protein affecting the JNK signaling pathway. CYLD is a negative regulator of TRAF2 and NF-kappa-B signaling pathway and is also known to have receptor-dependent role in regulating the I-kappa-B kinase pathway. It also has a deubiquitinating activity that is directed towards non-Lys-48-linked polyubiquitin chains and TRAP1 is a novel substrate for deubiquitination. Mutated CYLD is known to be associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome.
Specifications
| CYLD | |
| Polyclonal | |
| Unconjugated | |
| CYLD | |
| 2010013M14Rik; 2900009M21Rik; BRSS; C130039D01Rik; CDMT; Cyld; CYLD lysine 63 deubiquitinase; CYLD1; CYLDI; cylindromatosis (turban tumor syndrome); deubiquitinating enzyme CYLD; EAC; FLJ20180; FLJ31664; FLJ78684; HSPC057; KIAA0849; LRRGT00003; MFT; MFT1; mKIAA0849; probable ubiquitin carboxyl-terminal hydrolase CYLD; retinitis pigmentosa 1 homolog; Rp1; Rp1h; SBS; TEM; ubiquitin carboxyl-terminal hydrolase CYLD; ubiquitin specific peptidase like 2; ubiquitin thioesterase CYLD; ubiquitin thiolesterase CYLD; ubiquitin-specific-processing protease CYLD; Unknown (protein for MGC:137952); USPL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQC7 | |
| CYLD | |
| Recombinant protein corresponding to Human CYLD. Recombinant protein control fragment (Product #RP-98337). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction