missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYLC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18729-1-AP-150UL
This item is not returnable.
View return policy
Description
Possible architectural role during spermatogenesis. May be involved in spermatid differentiation.Specifications
| CYLC2 | |
| Polyclonal | |
| Unconjugated | |
| CYLC2 | |
| CYL2, CYLC2, Cylicin 2, Cylicin II, Multiple band polypeptide II | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1539 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14093 | |
| CYLC2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering