missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYLC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567100
This item is not returnable.
View return policy
Description
Immunogen sequence: KKDSKKDDKKK DAKKNAESTE MESDLELKKD KKHSKEKKGS KKDIKKDARK DTESTDAEFD ESSKTGFKTS TKIKGSDTES EESLYKP Highest antigen sequence identity to the following orthologs - mouse 40%, rat 39%.
Possible architectural role during spermatogenesis. May be involved in spermatid differentiation.
Specifications
| CYLC1 | |
| Polyclonal | |
| Unconjugated | |
| CYLC1 | |
| CYCL1; CYL; CYL1; CYLC1; cylicin; cylicin 1; cylicin I; cylicin, basic protein of sperm head cytoskeleton 1; cylicin-1; Multiple-band polypeptide I; rCG38076-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1538 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35663 | |
| CYLC1 | |
| Recombinant Human CYLC1. Recombinant protein control fragment (Product #RP-107746). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction