missing translation for 'onlineSavingsMsg'
Learn More

Cyheptamide, TRC

Product Code. 30357589
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357589 Supplier Toronto Research Chemicals Supplier No. TRCC9890401G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyheptamide
CAS 7199-29-3
Molecular Formula C16 H15 N O
Quantity 1 g
Synonym 5H-Dibenzo[a,d]cycloheptene-5-carboxamide, 10,11-dihydro-, 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-carboxamide, AY 8682, BS 7029, Carbamazepam, Cyheptamide, ICI 51426
InChI Formula InChI=1S/C16H15NO/c17-16(18)15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8,15H,9-10H2,(H2,17,18)
SMILES NC(=O)C1c2ccccc2CCc3ccccc13
IUPAC Name 6,11-dihydro-5H-dibenzo[2,1-b:1',2'-e][7]annulene-11-carboxamide
Molecular Weight (g/mol) 237.3
Formula Weight 237.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.