missing translation for 'onlineSavingsMsg'
Learn More

(±)-Cyhalothric Acid, TRC

Product Code. 30368326
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368326 10 mg 10mg
30363335 25 mg -
30361299 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368326 Supplier Toronto Research Chemicals Supplier No. TRCC27241510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68127-59-3
Molecular Formula C9 H10 Cl F3 O2
Molecular Weight (g/mol) 242.6227
Synonym Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, [1α,3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (1R,3R)-rel- (9CI), rel-(1R,3R)-3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propen-1-yl]-2,2-dimethylcyclopropanecarboxylic acid, (±)-Cyhalothric acid, 3-((Z)-2-Chloro-3,3,3-trifluoroprop-1-enyl)-2,2-dimethylcyclopropanecarboxylic acid, 3-[(Z)-2-Chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropanecarboxylic acid, cis-3-[(Z)-2-Chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropanecarboxylic acid, cis-Cyhalothric acid
IUPAC Name (1R,3R)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(C)[C@@H](\C=C(/Cl)\C(F)(F)F)[C@H]1C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H10ClF3O2/c1-8(2)4(6(8)7(14)15)3-5(10)9(11,12)13/h3-4,6H,1-2H3,(H,14,15)/b5-3-/t4-,6-/m0/s1
Formula Weight 242.0321
Chemical Name or Material cis-Cyhalothric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.