missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyhalofop, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Cyhalofop (Cyhalofop acid), the primary metabolite of Cyhalofop-butyl (HY-B0861) in susceptible grasses, is the herbicidally active metabolite. Cyhalofop-butyl is an aryloxyphenoxypropionate post-emergence herbicide widely used around the world in agriculture.
Specifications
Specifications
| Chemical Name or Material | Cyhalofop |
| CAS | 122008-78-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H12FNO4 |
| Quantity | 10 mg |
| Synonym | Cyhalofop acid |
| Solubility Information | DMSO : ≥ 31 mg/mL (102.90 mM) |
| SMILES | C[C@@H](OC1=CC=C(OC2=CC=C(C#N)C=C2F)C=C1)C(O)=O |
| Molecular Weight (g/mol) | 301.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction