missing translation for 'onlineSavingsMsg'
Learn More

Cyhalofop, MedChemExpress

Product Code. 16483407
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16483407

Brand: Medchem Express HY17528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyhalofop (Cyhalofop acid), the primary metabolite of Cyhalofop-butyl (HY-B0861) in susceptible grasses, is the herbicidally active metabolite. Cyhalofop-butyl is an aryloxyphenoxypropionate post-emergence herbicide widely used around the world in agriculture.

Cyhalofop (Cyhalofop acid), the primary metabolite of Cyhalofop-butyl (HY-B0861) in susceptible grasses, is the herbicidally active metabolite. Cyhalofop-butyl is an aryloxyphenoxypropionate post-emergence herbicide widely used around the world in agriculture.

Specifications

Chemical Name or Material Cyhalofop
CAS 122008-78-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H12FNO4
Quantity 50 mg
Synonym Cyhalofop acid
Solubility Information DMSO : ≥ 31 mg/mL (102.90 mM)
SMILES C[C@@H](OC1=CC=C(OC2=CC=C(C#N)C=C2F)C=C1)C(O)=O
Molecular Weight (g/mol) 301.27
Formula Weight 301.27
Percent Purity 95.4%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.