missing translation for 'onlineSavingsMsg'
Learn More

CYFIP1 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18425604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425604 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425604 Supplier Novus Biologicals Supplier No. NBP150028C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYFIP1 Polyclonal antibody specifically detects CYFIP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYFIP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cytoplasmic FMR1 interacting protein 1, cytoplasmic FMR1-interacting protein 1, FLJ45151, KIAA0068cytoplasmic FMRP interacting protein 1, P140SRA-1, selective hybridizing clone, SHYC, Specifically Rac1-associated protein 1, Sra-1, SRA1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1203 and 1253 of human Cytoplasmic FMR1 Interacting Protein 1 using the numbering given in entry NP_055423.1 (GeneID 23191).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23191
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.