missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress CycLuc1

Product Code. 16425376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16425376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16425376 Supplier MedChemExpress Supplier No. HY111653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CycLuc1 is a brain penetrant luciferase substrate.

Specifications

CAS 1247879-16-8
Solubility DMSO : ≥ 83.33 mg/mL (272.87 mM)
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Quantity 100 mg
Molecular Weight (g/mol) 305.38 g/mol
Product Type Dye Reagents
Purity or Quality Grade 96.77%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.