missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin C, TRC

Product Code. 30133571
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133571 50 mg 50mg
30003361 10 mg 10mg
30006028 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133571 Supplier Toronto Research Chemicals Supplier No. TRCC98890550MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59787-61-0
Molecular Formula C62 H111 N11 O13
Molecular Weight (g/mol) 1218.61
Synonym Ciclosporin C, 7-L-Threonine-cyclosporin A, Cyclosporin C, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-threonyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl]
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-[(1R)-1-hydroxyethyl]-33-[(E,1R,2R)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
SMILES C\C=C\C[C@@H](C)[C@@H](O)[C@@H]1N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C(=O)[C@@H](NC1=O)[C@@H](C)O)C(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C62H111N11O13/c1-25-26-27-39(14)52(76)51-56(80)66-49(42(17)74)60(84)67(18)32-47(75)68(19)43(28-33(2)3)55(79)65-48(37(10)11)61(85)69(20)44(29-34(4)5)54(78)63-40(15)53(77)64-41(16)57(81)70(21)45(30-35(6)7)58(82)71(22)46(31-36(8)9)59(83)72(23)50(38(12)13)62(86)73(51)24/h25-26,33-46,48-52,74,76H,27-32H2,1-24H3,(H,63,78)(H,64,77)(H,65,79)(H,66,80)/b26-25+/t39-,40+,41-,42-,43+,44+,45+,46+,48+,49+,50+,51+,52-/m1/s1
Formula Weight 1217.8363
Chemical Name or Material Cyclosporin C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.