missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin B, TRC

Product Code. 30010365
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010365 25 mg 25mg
30003264 50 mg 50mg
30007221 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010365 Supplier Toronto Research Chemicals Supplier No. TRCC98890325MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63775-95-1
Molecular Formula C61 H109 N11 O12
Molecular Weight (g/mol) 1188.58
Synonym Ciclosporin B, 7-L-Alanine-cyclosporin A, Cyclosporin B, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., Antibiotic S 7481F2, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-alanyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl]
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-33-[(E,1R,2R)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28,30-decamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
SMILES C\C=C\C[C@@H](C)[C@@H](O)[C@@H]1N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C(=O)[C@H](C)NC1=O)C(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C61H109N11O12/c1-25-26-27-39(14)51(74)50-55(78)64-41(16)56(79)66(18)32-47(73)67(19)43(28-33(2)3)54(77)65-48(37(10)11)60(83)68(20)44(29-34(4)5)53(76)62-40(15)52(75)63-42(17)57(80)69(21)45(30-35(6)7)58(81)70(22)46(31-36(8)9)59(82)71(23)49(38(12)13)61(84)72(50)24/h25-26,33-46,48-51,74H,27-32H2,1-24H3,(H,62,76)(H,63,75)(H,64,78)(H,65,77)/b26-25+/t39-,40+,41+,42-,43+,44+,45+,46+,48+,49+,50+,51-/m1/s1
Formula Weight 1187.8257
Chemical Name or Material Cyclosporin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.