missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclosporin A, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Cyclosporin A |
| CAS | 59865-13-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C62 H111 N11 O12 |
| Quantity | 25 mg |
| Synonym | 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., 1: PN: US6686454 SEQID: 1 claimed protein, 7: PN: WO2000002548 PAGE: 30 claimed protein, Abrammune, Antibiotic S 7481F1, Arpimune ME, CICPORAL, Cicloral, Cicloral (antibiotic), Ciclosporin, Ciclosporina Germed, Cipol N, Ciqorin, Consupren, CyclASol, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl], Cyclokat, Cyclosporin, Cyclosporine, Cyclosporine A, Cypol-N, Debio088, Equoral, Gengraf, Imusporin, NSC 290193, Neoplanta, Neoral, OL 27-400, Optimmune, Papilock Mini, Ramihyphin A, Restasis, S-Neoral, SDZ-OXL 400, Sandimmun, Sandimmun Neoral, Sandimmun Optoral, Sandimmune, Sandimmune Neoral, Sang-35, SangCyA, Sigmasporin Microoral, Zinograf ME, Ciclosporin, Cyclo[[(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)oct-6-enoyl]-l-2-aminobutanoyl-N-methylglycyl-N-methyl-l-leucyl-l-valyl-N-methyl-l-leucyl-l-alanyl-d-alanyl-N-methyl-l-leucyl-N-methyl-l-leucyl-N-methyl-l-valyl], ciclosporin A, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., Abrammune, Antibiotic S 7481F1, Arpimune ME, CICPORAL, Cicloral, Cicloral (antibiotic), Ciclosporina Germed, Cipol N, Consupren, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl], Cyclokat, Cyclosporin, Cyclosporine, Cyclosporine A, Debio088, Equoral, Gengraf, Imusporin, NSC 290193, Neoplanta, Neoral, OL 27-400, Optimmune, Papilock Mini, Ramihyphin A, Restasis, S-Neoral, SDZ-OXL 400, Sandimmun, Sandimmun Neoral, Sandimmun Optoral, Sandimmune, Sandimmune Neoral, Sang-35, SangCyA, Sigmasporin Microoral, Zinograf ME |
| InChI Formula | InChI=1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1 |
| SMILES | CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C |
| IUPAC Name | (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-ethyl-33-[(E,1R,2R)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone |
| Molecular Weight (g/mol) | 1202.61 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction