missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin A, 99+%, Thermo Scientific™

Cyclosporin A, CAS # 59865-13-3, is a compound that shows immunosuppressive, antifungal, antirheumatic, and dermatologic activities. It is also a calcineurin and enzyme inhibitor.

Brand:  Thermo Scientific Alfa Aesar J63191.03

133.00 GBP valid until 2026-07-03
BEST PRICE on Promo! Use promo code "27965" to get your promotional price.


Quantity 1g

 View more versions of this product

Product Code. 15475449

  • ONLINE EXCLUSIVE PRICE
    £190.00
    £171.65 / 1g
    Save £18.35
    9% Off
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum
Applications
• Cyclosporin A demonstrates immunosuppressive, antifungal, antirheumatic, and dermatologic activities
• In in vivo and in vitro studies it displays tumorigenicity
• This compound has also been shown to display anti-asthmatic and anticoronaviral activities
• It inhibits phosphoprotein phosphatase
• Cyclosporin A binds to the cellular protein cytophilin and inhibits the enzyme calcineurin
• Furthermore, it can induce apoptosis and inhibit angiogenesis induced by VEGF
• It exerts potent immunosuppressive activity on T immune cells, and causes reversible inhibition of immunocompetent lymphocytes in the G0- and G1-phase of the cell cycle
• It also suppresses the hepatitis C virus replication in cultured hepatocytes
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

59865-13-3
1202.64
PMATZTZNYRCHOR-IMVLJIQENA-N
132274082
CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
C62H111N11O12
MFCD00274558
Cyclosporine A; Antibiotic S 7481F1
30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone

Specifications

59865-13-3
1g
MFCD00274558
14,2752
Soluble in dimethyl sulfoxide and ethanol; Insoluble in water.
CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
1202.64
1202.61
Cyclosporin A
White
C62H111N11O12
3647785
Cyclosporine A; Antibiotic S 7481F1
PMATZTZNYRCHOR-IMVLJIQENA-N
30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone
132274082
≥99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.