missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyltriphenylphosphonium bromide, 98%

Product Code. 11446127
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446127 5 g 5g
11456127 25 g 25g
11466127 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446127 Supplier Thermo Scientific Alfa Aesar Supplier No. B25102.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 14114-05-7 | C21H20BrP | 383.27 g/mol

It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14114-05-7
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011872
InChI Key XMPWFKHMCNRJCL-UHFFFAOYSA-M
Synonym cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
PubChem CID 2723931
IUPAC Name cyclopropyl(triphenyl)phosphanium;bromide
SMILES [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 183°C to 186°C
Quantity 5 g
Beilstein 6413483
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 383.27
Percent Purity 98%
Chemical Name or Material Cyclopropyltriphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.