missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium bromide, 0.5M in MeTHF

Product Code. 15448037
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448037 100 mL 100mL
15438037 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448037 Supplier Thermo Scientific Alfa Aesar Supplier No. H51158.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
SMILES Br[Mg]C1CC1

Specifications

Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name bromo(cyclopropyl)magnesium
Formula Weight 145.28
Concentration or Composition (by Analyte or Components) 0.5M in MeTHF
Chemical Name or Material Cyclopropylmagnesium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.