missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 99%

Product Code. 11995191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995191 250 mL Glass bottle 250mL
10163250 10 mL Glass bottle 10mL
10205630 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11995191 Supplier Thermo Scientific Acros Supplier No. 154882500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-30-0 | C3H7N | 57.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.09
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -50.0°C
Color Colorless
Density 0.8240g/mL
Boiling Point 49.0°C to 50.0°C
Flash Point −25°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C3H5NH2
Refractive Index 1.4190 to 1.4220
Quantity 250 mL
Beilstein 12,3
Specific Gravity 0.824
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 57.09
Percent Purity 99%
Physical Form Volatile Liquid
Chemical Name or Material Cyclopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.