missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 98+%

Product Code. 11451498
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451498 250 g 250g
11431498 10 g 10g
11441498 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451498 Supplier Thermo Scientific Alfa Aesar Supplier No. A13844.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-30-0 | C3H7N | 57.096 g/mol

Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Solubility
Miscible with water, ethanol,ether and chloroform.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.096
MDL Number MFCD00001301
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -10°C
Density 0.824
Boiling Point 49°C to 50°C
Flash Point −25°C (−13°F)
Refractive Index 1.4206
Quantity 250 g
UN Number UN2733
Beilstein 741858
Sensitivity Hygroscopic
Solubility Information Miscible with water,ethanol,ether and chloroform.
Formula Weight 57.1
Percent Purity ≥98%
Chemical Name or Material Cyclopropylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.