missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylamine, 98+%

Product Code. 11441498
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11441498

Brand: Thermo Scientific Alfa Aesar A13844.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-30-0 | C3H7N | 57.096 g/mol

Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylamine) is used in the preparation of N-[4-(4-fluoro)phenyl-2-aminothiazol-5-yl]pyrimidin-2-yl-alkylamine derivatives. It is also used to prepare Pt(CPA)2(bismethylthiomethylenepropanedioate) and Pt(CPA)2(bisethylthiomethylenepropanedioate) complexes.

Solubility
Miscible with water, ethanol,ether and chloroform.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-30-0
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.096
MDL Number MFCD00001301
InChI Key HTJDQJBWANPRPF-UHFFFAOYSA-N
Synonym cyclopropylamine, aminocyclopropane, cyclopropyl amine, unii-8pr8xth1x1, 8pr8xth1x1, cyclopropanamin, cyclopropyamine, cyclopropylamin, cylopropylamine, cycloproplyamine
PubChem CID 69828
ChEBI CHEBI:34660
IUPAC Name cyclopropanamine
SMILES C1CC1N

Specifications

Melting Point -10°C
Density 0.824
Boiling Point 49°C to 50°C
Flash Point −25°C (−13°F)
Refractive Index 1.4206
Quantity 50 g
UN Number UN2733
Beilstein 741858
Sensitivity Hygroscopic
Solubility Information Miscible with water,ethanol,ether and chloroform.
Formula Weight 57.1
Percent Purity ≥98%
Chemical Name or Material Cyclopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropanamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cyclopropanamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.