missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetylene, TRC

Product Code. 30360554
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360554 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360554 Supplier Toronto Research Chemicals Supplier No. TRCC98893525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropylacetylene
CAS 6746-94-7
Molecular Formula C5H6
Quantity 25 g
Synonym Ethynylcyclopropane, Cyclopropylethyne
InChI Formula InChI=1S/C5H6/c1-2-5-3-4-5/h1,5H,3-4H2
SMILES C#CC1CC1
IUPAC Name ethynylcyclopropane
Molecular Weight (g/mol) 66.1
Formula Weight 66.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.