missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetonitrile, 97%, Thermo Scientific™

Product Code. 10823971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10823971 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10823971 Supplier Thermo Scientific Acros Supplier No. 435000050
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6542-60-5
Color Colorless to Yellow
Density 0.8780g/mL
Boiling Point 142°C to 144°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H7N
Refractive Index 1.4225 to 1.4245 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.878
Synonym cyclopropylacetonitrile, cyclopropaneacetonitrile, 2-cyclopropylethanenitrile, cyclopropyl acetonitrile, cyclopropyl-acetonitrile, acmc-20albr, cyanomethyl cyclopropane, ksc352m6t
InChI Key FAUQRRGKJKMEIW-UHFFFAOYSA-N
SMILES C1CC1CC#N
IUPAC Name 2-cyclopropylacetonitrile
Molecular Weight (g/mol) 81.12
PubChem CID 643447
Formula Weight 81.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclopropylacetonitrile
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropylacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.