Learn More
Cyclopropylacetonitrile, 97%
Description
Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.
Solubility
Miscible with water, methylene chloride and alcohols.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.89 |
| Boiling Point | 142°C to 144°C |
| Flash Point | 43°C (109°F) |
| Odor | Strong |
| Refractive Index | 1.4245 |
| Quantity | 25 g |
| UN Number | UN3275 |
| Beilstein | 2036964 |
| Solubility Information | Miscible with water,methylene chloride and alcohols. |
| Formula Weight | 81.12 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.