Learn More
Cyclopropyl(trimethylsilyl)acetylene, 97%
Description
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.
Solubility
Sparingly Soluble (0.069 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 83°C (80mmHg) |
| Quantity | 5 g |
| Flash Point | 25°C (77°F) |
| Refractive Index | 1.454 |
| UN Number | UN1993 |
| Solubility Information | Sparingly Soluble (0.069g/L) (25°C). |
| IUPAC Name | 2-cyclopropylethynyl(trimethyl)silane |
| Formula Weight | 138.28 |
| Percent Purity | 97% |
| Chemical Name or Material | Cyclopropyl(trimethylsilyl)acetylene |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.