missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl(trimethylsilyl)acetylene, +97%, ACROS Organics™

Product Code. 10540954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10540954 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10540954 Supplier Thermo Scientific Acros Supplier No. 387520010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 81166-84-9
Color Colorless to Yellow
Boiling Point 83.0°C (80.0 mmHg)
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Molecular Formula C8H14Si
Linear Formula C3H5CCSi(CH3)3
Refractive Index 1.4530 to 1.4550
MDL Number MFCD00671355
Quantity 1 g
Synonym cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane
InChI Key IYQSVLJDYLPMCX-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC1CC1
IUPAC Name (2-cyclopropylethynyl)trimethylsilane
Molecular Weight (g/mol) 138.29
PubChem CID 2758018
Formula Weight 138.28
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopropyl(trimethylsilyl)acetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.