missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl(trimethylsilyl)acetylene, 97%

Product Code. p-7035760
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15490358

Brand: Thermo Scientific Alfa Aesar H53487.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81166-84-9 | C8H14Si | 138.29 g/mol

Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl(trimethylsilyl)acetylene is used in several addition reactions.

Solubility
Sparingly Soluble (0.069 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81166-84-9
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.29
MDL Number MFCD00671355
InChI Key IYQSVLJDYLPMCX-UHFFFAOYSA-N
Synonym cyclopropyl trimethylsilyl acetylene, cyclopropylethynyl trimethylsilane, 2-cyclopropylethynyl trimethylsilane, cyclopropylethynyl trimethyl silane, silane, cyclopropylethynyl trimethyl, cyclopropane, 2-trimethylsilyl ethynyl, silane, cyclopropylethynyl trimethyl-9ci, timtec-bb sbb009013, trimethylsilylethynylcyclopropane, cyclopropylethynyl-trimethylsilane
PubChem CID 2758018
SMILES C[Si](C)(C)C#CC1CC1

Specifications

Boiling Point 83°C (80mmHg)
Quantity 5 g
UN Number UN1993
Flash Point 25°C (77°F)
Solubility Information Sparingly Soluble (0.069g/L) (25°C).
IUPAC Name 2-cyclopropylethynyl(trimethyl)silane
Formula Weight 138.28
Percent Purity 97%
Refractive Index 1.454
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.