missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Carbinol, 99%, Thermo Scientific Chemicals

Catalog Number 10511341
Click to view available options
Quantity:
25 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
2516-33-8
C4H8O
72.11
MFCD00001309
GUDMZGLFZNLYEY-UHFFFAOYSA-N
cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol
75644
cyclopropylmethanol
OCC1CC1
This item is not returnable. View return policy
2516-33-8
C4H8O
72.11
MFCD00001309
GUDMZGLFZNLYEY-UHFFFAOYSA-N
cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol
75644
cyclopropylmethanol
OCC1CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -60°C
Color Colorless
Density 0.8900g/mL
Boiling Point 123°C to 124°C (738.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5CH2OH
Refractive Index 1.4310 to 1.433
Quantity 25 mL
Beilstein 06,4
Specific Gravity 0.89
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol,slightly soluble in carbon tetrachloride
Viscosity 4.04 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclopropyl carbinol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'