missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl bromide, 99%

Product Code. 10784021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10784021 25 mL Glass bottle 25mL
10447870 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10784021 Supplier Thermo Scientific Acros Supplier No. 111640250
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4333-56-6 | C3H5Br | 120.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4333-56-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00001271
InChI Key LKXYJYDRLBPHRS-UHFFFAOYSA-N
Synonym cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
PubChem CID 78037
IUPAC Name bromocyclopropane
SMILES C1CC1Br

Specifications

Color Colorless
Density 1.5100g/mL
Flash Point -7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5Br
Refractive Index 1.4590 to 1.461
Quantity 25 mL
Specific Gravity 1.51
Solubility Information Solubility in water: immiscible
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclopropyl bromide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopropyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.