missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic acid, 98%

Product Code. 11452307
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452307 250 g 250g
11442307 50 g 50g
11432307 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452307 Supplier Thermo Scientific Alfa Aesar Supplier No. A14215.30
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1759-53-1 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1759-53-1
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00001287
InChI Key YMGUBTXCNDTFJI-UHFFFAOYSA-N
Synonym carboxycyclopropane, cyclopropylcarboxylic acid, cyclopropionic acid, cyclopropane carboxylic acid, cyclopropanecarboxylicacid, cpc-acid, cyclopropyl carboxylic acid, trimethylenecarboxylic acid, cyclopropane carboylic acid, cyclopropane-carboxylic acid
PubChem CID 15655
ChEBI CHEBI:23500
IUPAC Name cyclopropanecarboxylic acid
SMILES C1CC1C(=O)O

Specifications

Melting Point 5°C
Density 1.086
Boiling Point 182°C to 184°C
Flash Point 71°C (159°F)
Refractive Index 1.439
Quantity 250 g
UN Number UN3265
Beilstein 969839
Formula Weight 86.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.