missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxaldehyde, 98%

Product Code. 11313736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313736 1 g 1g
11323736 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313736 Supplier Thermo Scientific Alfa Aesar Supplier No. L14931.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1489-69-6 | C4H6O | 70.091 g/mol

Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-69-6
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00012261
InChI Key JMYVMOUINOAAPA-UHFFFAOYSA-N
Synonym cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
PubChem CID 123114
IUPAC Name cyclopropanecarbaldehyde
SMILES C1CC1C=O

Specifications

Density 0.938
Boiling Point 98°C to 101°C
Flash Point 7°C (44°F)
Odor Stench
Refractive Index 1.4298
Quantity 1 g
UN Number UN2924
Beilstein 906781
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.