missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 366820010

Code : 29

Additional Details : CAS Number : 1489-69-6 Weight : 0.05100kg Transport : UN number : 1989 Chem class : 3 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10170322

  • £26.20 / 1g


EU Stock 16
Add to basket


Specifications

Chemical Identifiers

Cyclopropanecarboxaldehyde
1489-69-6
C4H6O
MFCD00012261
98%
98%
70.09

Specifications

Cyclopropanecarboxaldehyde
7°C
1g
1489-69-6
98%
1.4285 to 1.4305
C3H5CHO
70.09
Solubility in water: insoluble.
0.9380g/mL
98.0° to 101.0°C
98%
Glass bottle
98%
C4H6O
0.938
MFCD00012261
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclopropanecarboxaldehyde

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS